CH6371382009 Chem Impex International, Inc.

alpha-Methyl-L-4-Fluorophenylalanine

  • MDL number MFCD08061614
  • Cas Number : 130855-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/alpha-methyl-l-4-fluorophenylalanine/7877?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/alpha-methyl-l-4-fluorophenylalanine/7877?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart