CH6371382012 Chem Impex International, Inc.

Fmoc-alpha-methyl-D-4-fluorophenylalanine

  • MDL number MFCD08061622
  • Cas Number : 1217777-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66-72 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to light yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-alpha-methyl-d-4-fluorophenylalanine/7880?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-alpha-methyl-d-4-fluorophenylalanine/7880?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart