CH6371382013 Chem Impex International, Inc.

Fmoc-alpha-methyl-L-4-Fluorophenylalanine

  • MDL number MFCD08061624
  • Cas Number : 1175838-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-alpha-methyl-l-4-fluorophenylalanine/7881?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-alpha-methyl-l-4-fluorophenylalanine/7881?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart