CH6371382014 Chem Impex International, Inc.

D-2-Amino-4-bromo-4-pentenoic acid

  • MDL number MFCD06659108
  • Cas Number : 264903-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242-248 °C (dec.)
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Yellowish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-2-amino-4-bromo-4-pentenoic-acid/7882?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-2-amino-4-bromo-4-pentenoic-acid/7882?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart