CH6371382016 Chem Impex International, Inc.

Boc-D-2-amino-4-bromo-4-pentenoic acid

  • MDL number MFCD01320883
  • Cas Number : 149930-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-108 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-2-amino-4-bromo-4-pentenoic-acid/7884?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-2-amino-4-bromo-4-pentenoic-acid/7884?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart