CH6371382017 Chem Impex International, Inc.

Boc-L-2-amino-4-bromo-4-pentenoic acid

  • MDL number MFCD01320852
  • Cas Number : 151215-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-(S)-(2-bromoallyl)glycine
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-2-amino-4-bromo-4-pentenoic-acid/7885?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-2-amino-4-bromo-4-pentenoic-acid/7885?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart