CH6371382018 Chem Impex International, Inc.

Fmoc-D-2-amino-4-bromo-4-pentenoic acid

  • MDL number MFCD01311777
  • Cas Number : 220497-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Yellowish solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-2-amino-4-bromo-4-pentenoic-acid/7886?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-2-amino-4-bromo-4-pentenoic-acid/7886?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart