CH6371382020 Chem Impex International, Inc.

N-Boc-3-(S)-amino-2-(S)-hydroxy-3-naphthalen-2-yl-propionic acid

  • MDL number MFCD07363734
  • Cas Number : 959583-98-3
  • Molecular Weight : 331.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-106 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white to yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-boc-3-i-s-i-amino-2-i-s-i-hydroxy-3-naphthalen-2-yl-propionic-acid/7892?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-boc-3-i-s-i-amino-2-i-s-i-hydroxy-3-naphthalen-2-yl-propionic-acid/7892?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart