CH6371382022 Chem Impex International, Inc.

D-2-Carbamoylphenylalanine

  • MDL number MFCD06659103
  • Cas Number : 1217613-52-9
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Almost white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-2-carbamoylphenylalanine/7910?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-2-carbamoylphenylalanine/7910?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart