CH6371382024 Chem Impex International, Inc.

D-4-Carbamoylphenylalanine

  • MDL number MFCD06659105
  • Cas Number : 1217609-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225 ° (dec.)
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-4-carbamoylphenylalanine/7912?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-4-carbamoylphenylalanine/7912?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart