CH6371382025 Chem Impex International, Inc.

L-2-Carbamoylphenylalanine

  • MDL number MFCD06659112
  • Cas Number : 959581-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203-209 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-carbamoylphenylalanine/7913?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-carbamoylphenylalanine/7913?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart