CH6371382026 Chem Impex International, Inc.

L-3-Carbamoylphenylalanine

  • MDL number MFCD06659113
  • Cas Number : 1217651-22-3
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255-263 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Slight red to white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-3-carbamoylphenylalanine/7914?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-3-carbamoylphenylalanine/7914?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart