CH6371382029 Chem Impex International, Inc.

Boc-D-3-carbamoylphenylalanine

  • MDL number MFCD06659120
  • Cas Number : 1213190-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 150 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form Almost white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-3-carbamoylphenylalanine/7917?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-3-carbamoylphenylalanine/7917?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart