CH6371382030 Chem Impex International, Inc.

Boc-D-4-carbamoylphenylalanine

  • MDL number MFCD06659121
  • Cas Number : 1213639-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 300 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-4-carbamoylphenylalanine/7918?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-4-carbamoylphenylalanine/7918?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart