CH6371382032 Chem Impex International, Inc.

Boc-L-3-carbamoylphenylalanine

  • MDL number MFCD06659129
  • Cas Number : 943449-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-3-carbamoylphenylalanine/7920?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-3-carbamoylphenylalanine/7920?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart