CH6371382033 Chem Impex International, Inc.

Boc-L-4-carbamoylphenylalanine

  • MDL number MFCD01317030
  • Cas Number : 205126-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 303-307 °C (dec.)
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-4-carbamoylphenylalanine/7921?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-4-carbamoylphenylalanine/7921?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart