CH6371382046 Chem Impex International, Inc.

D-2,4-Dimethylphenylalanine

  • MDL number MFCD06659101
  • Cas Number : 465500-97-4
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to pale yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-2-4-dimethylphenylalanine/7939?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-2-4-dimethylphenylalanine/7939?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart