CH6371382047 Chem Impex International, Inc.

L-2,4-Dimethylphenylalanine

  • MDL number MFCD06659111
  • Cas Number : 259726-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 273-279 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-4-dimethylphenylalanine/7940?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-4-dimethylphenylalanine/7940?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart