CH6371382048 Chem Impex International, Inc.

Boc-D-2,4-dimethylphenylalanine

  • MDL number MFCD06659117
  • Cas Number : 791625-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Yellowish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-2-4-dimethylphenylalanine/7941?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-2-4-dimethylphenylalanine/7941?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart