CH6371382058 Chem Impex International, Inc.

threo-N-Boc-D-phenylalanine epoxide

  • MDL number MFCD08061626
  • Cas Number : 98818-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-threo-i-n-boc-d-phenylalanine-epoxide/7951?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-threo-i-n-boc-d-phenylalanine-epoxide/7951?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart