CH6371382059 Chem Impex International, Inc.

erythro-N-Boc-D-phenylalanine epoxide

  • MDL number MFCD08061627
  • Cas Number : 98818-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-erythro-i-n-boc-d-phenylalanine-epoxide/7952?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-erythro-i-n-boc-d-phenylalanine-epoxide/7952?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart