CH6371382073 Chem Impex International, Inc.

Fmoc-(S)-2-methoxyphenylglycine

  • MDL number MFCD07371754
  • Cas Number : 1176648-58-0
  • Molecular Weight : 403.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-2-methoxyphenylglycine/7966?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-2-methoxyphenylglycine/7966?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart