CH6371382077 Chem Impex International, Inc.

Boc-L-2,4,5-trifluorophenylalanine

  • MDL number MFCD06659127
  • Cas Number : 324028-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-124 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-2-4-5-trifluorophenylalanine/7970?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-2-4-5-trifluorophenylalanine/7970?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart