CH6371382080 Chem Impex International, Inc.

Fmoc-(R)-3-amino-4-(2,4,5-trifluorophenyl)-butyric acid

  • MDL number MFCD06659148
  • Cas Number : 1217818-53-5
  • Molecular Weight : 455.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-4-2-4-5-trifluorophenyl-butyric/7973?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-r-i-3-amino-4-2-4-5-trifluorophenyl-butyric/7973?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart