CH6371382081 Chem Impex International, Inc.

(S)-3-Amino-4-(2,4,5-trifluorophenyl)butyric acid HCl

  • MDL number MFCD06659149
  • Cas Number : 1217809-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-222 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-4-2-4-5-trifluorophenyl-butyric-acid-font-size-2-face-symbol-middot-font-hcl/7974?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-3-amino-4-2-4-5-trifluorophenyl-butyric-acid-font-size-2-face-symbol-middot-font-hcl/7974?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart