CH6371382083 Chem Impex International, Inc.

Fmoc-(S)-3-amino-4-(2,4,5-trifluorophenyl)butyric acid

  • MDL number MFCD06659151
  • Cas Number : 959580-94-0
  • Molecular Weight : 455.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-4-2-4-5-trifluorophenyl-butyric-acid/7976?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-3-amino-4-2-4-5-trifluorophenyl-butyric-acid/7976?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart