CH6371382095 Chem Impex International, Inc.

Boc-2-(Fmoc-aminomethyl)-L-phenylalanine

  • MDL number MFCD06659130
  • Cas Number : 959573-16-1
  • Molecular Weight : 516.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-fmoc-aminomethyl-l-phenylalanine/7989?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-fmoc-aminomethyl-l-phenylalanine/7989?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart