CH6371382102 Chem Impex International, Inc.

Fmoc-3-(Boc-aminomethyl)-D-phenylalanine

  • MDL number MFCD03093567
  • Cas Number : 1217665-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-boc-aminomethyl-d-phenylalanine/7996?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-boc-aminomethyl-d-phenylalanine/7996?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart