CH6371382105 Chem Impex International, Inc.

Boc-(2S-3S)-3-amino-2-hydroxy-4-phenylbutyric acid

  • MDL number MFCD00829437
  • Cas Number : 116661-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-2-i-s-i-3-i-s-i-3-amino-2-hydroxy-4-phenylbutyric-acid/7999?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-2-i-s-i-3-i-s-i-3-amino-2-hydroxy-4-phenylbutyric-acid/7999?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart