CH6371382106 Chem Impex International, Inc.

(S)-alpha-Benzylproline HCl

  • MDL number MFCD06659152
  • Cas Number : 637020-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white to yellowish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-a-benzylproline-font-size-2-face-symbol-middot-font-hcl/8000?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-a-benzylproline-font-size-2-face-symbol-middot-font-hcl/8000?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart