CH6371382108 Chem Impex International, Inc.

(2R,3R)-3-Amino-2-hydroxy-3-phenyl-propionic acid

  • MDL number MFCD07363513
  • Cas Number : 55325-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-222°C (Dec.)
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-i-r-i-3-i-r-i-3-amino-2-hydroxy-3-phenyl-propionic-acid/8003?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-i-r-i-3-i-r-i-3-amino-2-hydroxy-3-phenyl-propionic-acid/8003?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart