CH6371382114 Chem Impex International, Inc.

Fmoc-O-allyl-D-m-tyrosine

  • MDL number MFCD08061621
  • Cas Number : 1217835-37-4
  • Molecular Weight : 443.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-102 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-allyl-d-i-m-i-tyrosine/8010?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-allyl-d-i-m-i-tyrosine/8010?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart