CH6371382128 Chem Impex International, Inc.

Fmoc-(R,S)-3,4-cis-methanoproline

  • MDL number MFCD08064307
  • Cas Number : 1820598-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at = -4 °C
Physical Form White to pale yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-r-s-3-4-i-cis-i-methanoproline/8031?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-r-s-3-4-i-cis-i-methanoproline/8031?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart