CH6371382138 Chem Impex International, Inc.

Fmoc-Tyr(tBu)-Thr[Psi(Me,Me)Pro]-OH

  • MDL number MFCD08064321
  • Cas Number : 920519-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95101
Storage temp. Store at = -4 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-tyr-tbu-thr-psi-sup-me-me-sup-pro-oh/8046?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-tyr-tbu-thr-psi-sup-me-me-sup-pro-oh/8046?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart