CH6371382216 Chem Impex International, Inc.

4-(2,4-Difluorobenzoyl)piperidine hydrochloride

  • MDL number MFCD01313310
  • Cas Number : 106266-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2,4-Difluorophenyl)piperidin-4-yl-methanone hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white to light yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-2-4-difluorobenzoyl-piperidine-hydrochloride/8125?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-2-4-difluorobenzoyl-piperidine-hydrochloride/8125?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart