CH6371382220 Chem Impex International, Inc.

2-(3,5-Dimethylpyrazol-1-yl)ethylamine

  • MDL number MFCD04967722
  • Cas Number : 62821-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Clear colorless liquid
Shipping Temperature RT
Country of Origin USA
Boiling point 108-114 °C (12mm)
Product Link http://www.chemimpex.com/product/productinfo/2-3-5-dimethylpyrazol-1-yl-ethylamine/8129?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-3-5-dimethylpyrazol-1-yl-ethylamine/8129?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart