CH6371382342 Chem Impex International, Inc.

2-(5-Chloro-1H-benzoimidazol-2-yl)ethylamine dihydrochloride

  • MDL number MFCD02255631
  • Cas Number : 135875-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8°C
Physical Form Brown solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-5-chloro-1-i-h-i-benzoimidazol-2-yl-ethylamine/8256?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-5-chloro-1-i-h-i-benzoimidazol-2-yl-ethylamine/8256?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart