CH6371382393 Chem Impex International, Inc.

2-(2H-Tetrazol-5-yl)pyridine

  • MDL number MFCD00269436
  • Cas Number : 33893-89-9
  • Molecular Weight : 147.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216 - 220 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White needles
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-2h-tetrazol-5-yl-pyridine/8313?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-2h-tetrazol-5-yl-pyridine/8313?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart