CH6371382406 Chem Impex International, Inc.

2-Piperidin-3-yl-1,3-benzoxazole, free base

  • MDL number MFCD06660810
  • Cas Number : 51785-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Clear, viscous yellow liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-piperidin-3-yl-1-3-benzoxazole-free-base/8329?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-piperidin-3-yl-1-3-benzoxazole-free-base/8329?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart