CH6371382426 Chem Impex International, Inc.

5-(4-Chlorobenzyl)-2H-tetrazole

  • MDL number MFCD00205200
  • Cas Number : 14064-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161 - 166 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White needles
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-chlorobenzyl-2-i-h-i-tetrazole/8353?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-chlorobenzyl-2-i-h-i-tetrazole/8353?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart