CH6371382453 Chem Impex International, Inc.

3,4-Dimethoxybenzophenone

  • MDL number MFCD00075875
  • Cas Number : 4038-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-57 �C
Synonym (3,4-Dimethoxyphenyl)phenyl-methanone
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-4-dimethoxybenzophenone/8386?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-4-dimethoxybenzophenone/8386?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart