CH6371382534 Chem Impex International, Inc.

2,4-Dichloro-1-[isocyano-(toluene-4-sulfonyl)methyl]benzene

  • MDL number MFCD04114807
  • Cas Number : 952727-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Colorless to yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-4-dichloro-1-isocyano-toluene-4-sulfonyl-methyl-benzene/8485?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-4-dichloro-1-isocyano-toluene-4-sulfonyl-methyl-benzene/8485?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart