CH6371382549 Chem Impex International, Inc.

N-Methyl-4-piperidinecarboxylic acid methyl ester

  • MDL number MFCD06658483
  • Cas Number : 1690-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 1-methylpiperidine-4-carboxylate
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-4-piperidinecarboxylic-acid-methyl-ester/8502?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-4-piperidinecarboxylic-acid-methyl-ester/8502?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart