CH6371382562 Chem Impex International, Inc.

2-Chloro-Imidazo[1,2-a]pyridine-3-carbaldehyde

  • MDL number MFCD02708012
  • Cas Number : 131773-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Brown crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-chloro-imidazo-1-2-a-pyridine-3-carbaldehyde/8522?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-chloro-imidazo-1-2-a-pyridine-3-carbaldehyde/8522?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart