CH6371382601 Chem Impex International, Inc.

4-Methylpyridin-2-yl boronic acid

  • MDL number MFCD03093330
  • Cas Number : 372963-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methylpyridin-2-yl-boronic-acid/8566?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methylpyridin-2-yl-boronic-acid/8566?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart