CH6371382636 Chem Impex International, Inc.

4-(4-Methoxyphenyl)pyrrolidine-3-methylcarboxylate

  • MDL number MFCD01862548
  • Cas Number : 939758-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-4-methoxyphenyl-pyrrolidine-3-methylcarboxylate/8609?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-4-methoxyphenyl-pyrrolidine-3-methylcarboxylate/8609?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart