CH6371382696 Chem Impex International, Inc.

2-(2-Allyloxy-phenoxymethyl)oxirane

  • MDL number MFCD06658531
  • Cas Number : 6452-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Pale oily liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-2-allyloxy-phenoxymethyl-oxirane/8692?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-2-allyloxy-phenoxymethyl-oxirane/8692?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart