CH6371382719 Chem Impex International, Inc.

2-Iodo-3-methoxypyridine

  • MDL number MFCD00234181
  • Cas Number : 93560-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-iodo-3-methoxypyridine/8733?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-iodo-3-methoxypyridine/8733?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart