CH6371382740 Chem Impex International, Inc.

3-Methylbenzenecarboximidamide hydrochloride

  • MDL number MFCD02180878
  • Cas Number : 20680-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Methyl-benzamidine hydrochloride
Purity 95
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-methylbenzenecarboximidamide-hydrochloride/8758?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-methylbenzenecarboximidamide-hydrochloride/8758?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart