CH6371382753 Chem Impex International, Inc.

3-Carboxymethyl-morpholine-4-carboxylic acid tert-butyl ester

  • MDL number MFCD04114891
  • Cas Number : 859155-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-carboxymethyl-morpholine-4-carboxylic-acid-i-tert-i-butyl-ester/8774?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-carboxymethyl-morpholine-4-carboxylic-acid-i-tert-i-butyl-ester/8774?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart